Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BMP10 Rabbit Polyclonal Antibody

Supplier: BioVision

5581-100
10008-092EA 252.35 USD InStock
10008-092
Anti-BMP10 Rabbit Polyclonal Antibody
Antibodies
BMPs (bone morphogenetic proteins) belong to the TGF-β superfamily of structurally related signaling proteins. As implied by their name, BMPs promote and regulate bone development, growth, remodeling and repair, in both prenatal development and postnatal growth of eye, heart, kidney, skin, and other tissues. BMP-10 is essential for the proper development of the heart, but is not expressed until after cardiac patterning or looping is completed.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BMP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty