Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PAI-1 Rabbit Polyclonal Antibody

Supplier: BioVision

5579-100
10007-854EA 272.95 USD InStock
10007-854
Anti-PAI-1 Rabbit Polyclonal Antibody
Antibodies
PAI-1 (plasminogen activator inhibitor-1) is originally cloned from human endothelial cell and rat hepatoma cell cDNA libraries. As a member of the serpin family of serine protease inhibitors, PAI-1 inhibits both tissue-type palsminogen activator (t-PA) and urokinase-type plasminogen activator (u-PA). High PAI-1 levels are associated with an increased risk of thromboembolic disease while PAI-1 deficiency may represent an inherited autosomal recessive bleeding disorder.

Recommended Dilutions: Western Blot: 1-2 µg/ml

Type: Primary
Antigen: PAI-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty