Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DAP12 Rabbit Polyclonal Antibody

Supplier: BioVision

5570R-100
10007-840EA 288.4 USD InStock
10007-840
Anti-DAP12 Rabbit Polyclonal Antibody
Antibodies
DAP12 (KARAP) is a natural killer (NK) cell-expressed cell surface receptor and belongs to the immunoglobulin and C-type lectin superfamily. Unlike other members of this family which possess immunoreceptor tyrosine-based inhibitory motifs (ITIM), DAP12 contains an immunoreceptor tyrosine-based activation motif (ITAM) in its cytoplasmic domain and is present as a disulphide-bonded homodimer. Crosslinking of DAP 12-KIR complexes (membrane glycoproteins of the killer-cell inhibitory receptor family without an ITIM in their cytoplasmic domain) results in cellular activation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DAP12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty