Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMP9 Rabbit Polyclonal Antibody

Supplier: BioVision

5565-100
10007-838EA 272.95 USD InStock
10007-838
Anti-MMP9 Rabbit Polyclonal Antibody
Antibodies
The mammalian Matrix metalloproteinases (MMPs) degrade extracellular matrix in physiological and pathological processes. After cleavage of a single peptide domain of about 20 amino acids, the MMPs are secreted in latent forms. Upon activation, the N-terminal propeptide domain is cleaved to generate the active forms of MMP. MMP-9 (92 kDa type IV collagenase, Gelatinase-B) contains the basic structure of propeptide, catalytic, and hemopexin domains. It is an important proteinase in tissue remodeling.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 4-8 µg/ml; and Immunohistochemistry: 20-40 µg/ml

Type: Primary
Antigen: MMP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty