Anti-MMP2 Rabbit Polyclonal Antibody

Supplier: BioVision
5562-100
10007-834EA 346.02 USD
10007-834
Anti-MMP2 Rabbit Polyclonal Antibody
Antibodies
The mammalian Matrix metalloproteinases (MMPs) degrade extracellular matrix in physiological and pathological processes. After cleavage of a single peptide domain of about 20 amino acids, the MMPs are secreted in latent forms. Upon activation, the N-terminal propeptide domain is cleaved to generate the active forms of MMP. MMP-2 (72 kDa type IV collagenase, Gelatinase-A) contains the basic structure of propeptide, catalytic, and hemopexin domains. It is an important proteinase in tissue remodeling.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 4-8 µg/ml

Type: Primary
Antigen: MMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR