Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MKK3 Rabbit Polyclonal Antibody

Supplier: BioVision

3285-100
10003-750EA 272.95 USD InStock
10003-750
Anti-MKK3 Rabbit Polyclonal Antibody
Antibodies
MKK3 is a protein kinase that phosphorylates the p38 MAPK. Phosphorylation by MKK3 occurs on threonine and tyrosine residues and increases the activity of p38 to stimulate transcription factors ATF2 and Elk-1. MKK3, together with MKK6, serves as upstream regulators of p38 MAPK activation. A structural variant, MKK3b, has been identified that contains 29 more amino acids at its N-terminus.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; immunoprecipitation

Type: Primary
Antigen: MKK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty