Anti-CASP12 Rabbit Polyclonal Antibody

Supplier: BioVision
3282-100
10003-744EA 326.9 USD
10003-744
Anti-CASP12 Rabbit Polyclonal Antibody
Three distinct signaling pathways lead to programmed cell death (apoptosis). The death receptor and mitochondrion pathways are the mains two pathways, in which the key apoptotic proteases caspase-8 and caspase-9, respectively, are involved. The endoplasmic reticulum (ER) stress is the third apoptotic pathway and caspase-12 is involved. Caspase-12 is located to the ER, but not to the cytoplasm or mitochondrion. Caspase-12 is activated by ER stress, including disruption of ER calcium hemeostasis, and mediates ER stress-induced apoptosis. Caspase-12 is localized to the ER with several proteins that are involved in Alzheimer’s disease including gamma-secretase presenilin and β-amyloid precursor protein (APP). Caspase-12 mediates cytotoxicity induced by amyloid-β. Caspase-12 is ubiquitously expressed in mouse tissues.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CASP12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR