Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MBD1 Rabbit Polyclonal Antibody

Supplier: BioVision

3281-100
10003-740EA 252.35 USD InStock
10003-740
Anti-MBD1 Rabbit Polyclonal Antibody
Antibodies
The MBD family of proteins includes MeCP2, MBD1, MBD2, MBD3 and MBD4. MBD1 (methyl-CpG binding domain protein 1) binds to methylated CpG sites and represses gene transcription. MBD1 is recruited to both methylated and non-methylated CpGs by separate domains: the CXXC domain targets the protein to non-methylated CpGs whereas the MBD domain targets the protein to methylated CpG. MBD1 may also interact with histone methyltransferase SETDB1 to repress transcription.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MBD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty