Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RGS3 Rabbit Polyclonal Antibody

Supplier: BioVision

3280-100
10003-738EA 272.95 USD InStock
10003-738
Anti-RGS3 Rabbit Polyclonal Antibody
Antibodies
RGS3 (regulator of G protein signaling 3) is a 519-residue protein that is widely expressed in many tissues and cells. The protein contains an RGS domain near the carboxy terminus that serves as a GTPase-activating protein (GAP). RGS3 negatively regulates the activation of Giα and Gαq. A truncated RGS3 (RGS3T), with its N-terminal 313 residues removed, has been shown to negatively regulate acenylyl cyclase and PLC. RGS3T may be localized in the nucleus and RGS3 is found in cytoplasm. Translocation to membrane is important for the RGS proteins to interact with heterotrimeric G proteins.

Recommended Dilutions: Western Blot: 1 µg/ml;

Type: Primary
Antigen: RGS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty