Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RIP Rabbit Polyclonal Antibody

Supplier: BioVision

3279-100
10003-734EA 262.65 USD InStock
10003-734
Anti-RIP Rabbit Polyclonal Antibody
Antibodies
RIP (Receptor Interacting Protein) is a 74 kDa Ser/Thr kinase which interacts with CD95 (Fas/Apo1) receptor and the tumor necrosis factor receptor (TNFR1). It is a cell death domain adapter protein which can bind to the adapter proteins TRADD, RAID, and TRAF2. RIP contains an N-terminal region with homology to protein kinase, an intermediate domain capable of association with MAPKKK and a C-terminal region containing an intracellular death domain motif. RIP activates both p38 MAP kinase and SAPK families. In vitro, RIP induces apoptosis, as well as SAPK/JNK and NF-kB activation. RIP possesses kinase activity as it autophosphorylates itself on serine and threonine residues.

Recommended Dilutions: Western Blot: 2-6 µg/ml;

Type: Primary
Antigen: RIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty