Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-JMJD1A Rabbit Polyclonal Antibody

Supplier: BioVision

3273-100
10003-720EA 252.35 USD InStock
10003-720
Anti-JMJD1A Rabbit Polyclonal Antibody
Antibodies
JMJD1A (Jumonji domain containing 1A), belongs to the JHDM2 histone demethylase family. Histone demethylase specifically demethylates 'Lys-9' of histone H3, thus play a central role in histone code. JMJD1A is involved in spermatogenesis by regulating expression of target genes such as PRM1 and TMP1 that are required for proper packaging and condensation of sperm chromatin.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: JMJD1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty