Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MBD2a Rabbit Polyclonal Antibody

Supplier: BioVision

3232-100
10003-590EA 252.35 USD InStock
10003-590
Anti-MBD2a Rabbit Polyclonal Antibody
Antibodies
The MBD family of proteins includes MeCP2, MBD1, MBD2, MBD3 and MBD4. MBD2 is highly similar to MBD3 but only MBD2 targets the MeCP1 complex to methylate DNA. MBD2 has two isoforms, MBD2a and MBD2b. MBD2a is a 414 amino acids protein, whereas MBD2b lacks the 152 amino acids N-terminal extension. MBD2a interacts with RNA helicase A to enhance CREB-dependent gene exptression.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MBD2a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty