Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DNMT3A Rabbit Polyclonal Antibody

Supplier: BioVision

3227-100
10003-576EA 283.25 USD InStock
10003-576
Anti-DNMT3A Rabbit Polyclonal Antibody
Antibodies
Methylation of DNA at cytosine residues plays an important role in regulation of gene expression, genomic imprinting, and is essential for mammalian development. Hypermethylation of CpG islands in tumor suppressor genes or hypomethylation of bulk genomic DNA may be linked with development of cancer. To date, three families of mammalian DNA methyltransferase genes have been identified which include DNMT1, DNMT2, and DNMT3. DNMT1 is constitutively expressed in proliferating cells and inactivation of this gene causes global demethylation of genomic DNA and embryonic lethality. DNMT2 is expressed at low levels in adult tissues and its inactivation does not affect DNA methylation or maintenance of methylation. The DNMT3 family members, DNMT3a and DNMT3b, are strongly expressed in embryonic stem (ES) cells but their expression is down regulated in differentiating ES cells and is low in adult somatic tissue. Recently, it has been shown that naturally occurring mutations of DNMT3b gene occur in patients with a rare autosomal recessive disorder, termed ICF (immunodeficiency, centromeric instability, and facial anomalies) syndrome.

Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: DNMT3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty