Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SLAM Rabbit Polyclonal Antibody

Supplier: BioVision

3226-200
10013-886EA 272.95 USD InStock
10013-886
Anti-SLAM Rabbit Polyclonal Antibody
Antibodies
Signaling lymphocytic activation molecule (SLAM/CD150) is expressed at hematopoietic tissue, such as memory T-cells, as well as at immature thymocytes. It has high-affinity self-ligand ability and plays an important role in interaction of T and B cells.

Recommended Dilutions: Western Blot: 3-6 mg/ml

Type: Primary
Antigen: SLAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Ig
Reactivity: Human, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty