Anti-SIRT5 Rabbit Polyclonal Antibody

Supplier: BioVision
3225-100
10013-882EA 286.04 USD
10013-882
Anti-SIRT5 Rabbit Polyclonal Antibody
Silent information regulator (Sir2)-like family deacetylases (also known as sirtuins) are highly conserved proteins and have important roles in the regulation of metabolism, inflammation, cellular survival growth and differentiation. SIRT5 is a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Sirtuins are NAD-dependent protein ADP-ribosyl transferase which catalyzes the transfer of ADP-ribosyl groups onto target proteins. Alternative splicing of this gene results in two transcript variants.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: SIRT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR