Anti-SIRT4 Rabbit Polyclonal Antibody

Supplier: BioVision
AB124521-100UG
10003-574EA 811.12 USD
10003-574
Anti-SIRT4 Rabbit Polyclonal Antibody
Antibodies
Silent information regulator (Sir2)-like family deacetylases (also known as sirtuins) are highly conserved proteins and have important roles in the regulation of metabolism, inflammation, cellular survival growth and differentiation. Sirtuins, including SIRT1-7, are human homologs of yeast Sir2p. Sirtuins are NAD-dependent protein ADP-ribosyl transferase which catalyzes the transfer of ADP-ribosyl groups onto target proteins, including mitochondrial GLUD1. SIRT4 localizes to mitochondria, inhibits glutamate dehydrogenase (GLUD1), and may involve in the regulation of insulin secretion.

Recommended Dilutions: Western Blot: 1-4 µg/ml

Type: Primary
Antigen: SIRT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR