Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SIRT3 Rabbit Polyclonal Antibody

Supplier: BioVision

3223-100
10003-570EA 252.35 USD InStock
10003-570
Anti-SIRT3 Rabbit Polyclonal Antibody
Antibodies
Silent information regulator (Sir2)-like family deacetylases (also known as sirtuins) are highly conserved proteins and have important roles in the regulation of metabolism, inflammation, cellular survival growth and differentiation. SIRT3 is a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Yeast sirtuin proteins are found to regulate epigenetic gene silencing. Sirtuins are NAD-dependent protein ADP-ribosyl transferase which catalyzes the transfer of ADP-ribosyl groups onto target proteins.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SIRT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty