Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DR6 Rabbit Polyclonal Antibody

Supplier: BioVision

3216R-100
10003-560EA 252.35 USD InStock
10003-560
Anti-DR6 Rabbit Polyclonal Antibody
Antibodies
Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family throμgh their death domain containing receptors, TNF-R1 and Fas. Several novel death receptor including DR3, DR4, and DR5 were recently identified. A new death domain containing receptor in the TNFR family was cloned recently and termed DR6 for death receptor 6. Like TNF-R1, DR6 interacts with death domain containing adapter molecule TRADD. Overexpression of DR6 induces apoptosis and activates NF-kB and JNK. DR6 is widely expressed in human tissues and cell lines.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: DR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty