Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Nestin Rabbit Polyclonal Antibody

Supplier: BioVision

3208-100
10003-556EA 293.55 USD InStock
10003-556
Anti-Nestin Rabbit Polyclonal Antibody
Antibodies
Nestin may play a role in the trafficking and distribution of IF proteins and potentially other cellular factors to daμghter cells during progenitor cell division (By similarity).

Recommended Dilutions: Western Blot: 1:100~500; ELISA: ~1:1,000; Flow Cytometry: 1:10~50).

Type: Primary
Antigen: Nestin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Ig
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty