Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GFAP Rabbit Polyclonal Antibody

Supplier: BioVision

3206-200
10003-552EA 283.25 USD InStock
10003-552
Anti-GFAP Rabbit Polyclonal Antibody
Antibodies
GFAP, Glial fibillary acidic protein is an intermediate filament protein. It was found in astrocytes cells as a cell specific marker in the central nervous system development. GFAP is defective in Alexander disease. But it is highly expressed in Astrogliosis which is a result of some diseases, such as AIDS, dementia and inflammatory demyelination diseases.

Recommended Dilutions: Western Blot: 4-10 µg/ml

Type: Primary
Antigen: GFAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty