Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Bnip3L/Nix Rabbit Polyclonal Antibody

Supplier: BioVision

3205-100
10003-550EA 252.35 USD InStock
10003-550
Anti-Bnip3L/Nix Rabbit Polyclonal Antibody
Antibodies
Members of Bcl-2 family are critical regulators of apoptosis by either inhibiting or promoting cell death. Bcl-2 homology 3 (BH3) domain is a potent death domain. BH3 domain containing pro-apoptotic proteins, including Bad, Bax, Bid, Bik, Hrk, Nip3, and Bim, form a growing subclass of the Bcl-2 family. A novel BH3 domain containing protein was recently identified and designated Bnip3L, Bnip3α, and Nix (for Nip3-like protein X). Bnip3L/Bnip3α/Nix is a homolog of the E1B 19K/Bcl-2 binding and pro-apoptotic protein Bnip3. Overexpression of Bnip3L induces apoptosis. Bnip3L interacts with and overcomes supression by Bcl-2 and Bcl-xL. Bnip3L is localized in mitochondria. The message RNA of Bnip3L and Bnip3 form a new subfamily of the proapoptotic mitochondrial proteins.

Recommended Dilutions: Western Blot: 0.5-2 µg/ml; Immunohistochemistry: 2-10 µg/ml

Type: Primary
Antigen: Bnip3L/Nix
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty