Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BAFF Rabbit Polyclonal Antibody

Supplier: BioVision

3203-100
10003-548EA 252.35 USD InStock
10003-548
Anti-BAFF Rabbit Polyclonal Antibody
Antibodies
BAFF (for B cell Activating Factor) is a novel member in the TNF family. BAFF/BLyS was characterized as a B cell stimulator since it induced B cell proliferation and immunoglobulin secretion. Two receptors for BAFF were recently identified and designated TACI and BCMA. BAFF and its receptors are involved in the development of systemic lupus erythaematosus and other B cell associated autoimmune diseases. Like TNF-α and TRAIL, THANK was shown to activate NF-kB and c-Jun N-terminal kinase (JNK) and to induce apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: BAFF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty