Anti-ADAM10 Rabbit Polyclonal Antibody

Supplier: BioVision
3201-100
10003-544EA 309.39 USD
10003-544
Anti-ADAM10 Rabbit Polyclonal Antibody
Proinflammatory cytokine tumor necrosis factorα (TNF-α) contributes to a variety of inflammatory responses and programmed cell death. Notch receptor and its ligand participate in cell fate decisions during vertebrate development and are associated with several human disorders, including a T-cell lymphoma. TNF-α, notch and its ligand delta are all membrane-bond molecules, which are cleaved by proteases to release mature proteins or functional receptor. ADAM10, a metalloprotease-disintegrin in the family of mammalian ADAM (for a disintegrin and metalloprotease), was recently identified to cleave TNF-α, notch and its ligand delta. The genes encoding human, mouse, and bovine ADAM10 were recently cloned and designated ADAM10, kuzbanian (KUZ), and MADM, respectively. ADAM10 mRNA is expressed in a variety of human and bovine tissues.

  • Recommended Dilutions: Western Blot: 1-4 µg/ml; Immunofluorescence: 2-10 µg/ml

Type: Primary
Antigen: ADAM10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR