Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TGF-beta1 Rabbit Polyclonal Antibody

Supplier: BioVision

5559-100
10007-828EA 319.3 USD InStock
10007-828
Anti-TGF-beta1 Rabbit Polyclonal Antibody
Antibodies
TGF-β(transforming growth factor-beta) is a family of multifunctional proteins. TGF-β was originally identified for its ability to induce the growth of normal rodent fibroblast in soft agar. It is now known that TGF-β is a potent growth inhibitor of many normal and transformed cell lines. It regulates normal cell growth, development and tissue remodeling following injury. TGF-β is produced as latent high molecular weight complexes. The activation of latent TGF-β is an important step in the regulation of its action.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: TGF-beta1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Pig

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty