Anti-sRANK Ligand Polyclonal Rabbit Polyclonal Antibody

Supplier: BioVision
5557-100
10007-826EA 288.13 USD
10007-826
Anti-sRANK Ligand Polyclonal Rabbit Polyclonal Antibody
RANK (receptor activator of NF-kB) is a member of the TNFR family, identified as a dendritic cell membrane protein. Murine sRANK Ligand is a soluble 19.4 kDa protein, comprising the TNF-like extracellular domain of RANKL (174 amino acids). The full length of RANK Ligand is 39.6 kDa containing 36 amino acids.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: sRANK Ligand Polyclonal
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR