Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL21R Rabbit Polyclonal Antibody

Supplier: BioVision

5554-100
10007-824EA 272.95 USD InStock
10007-824
Anti-IL21R Rabbit Polyclonal Antibody
Antibodies
IL-21 is a novel cytokine that was recently identified in human and mouse. It has significant homology to IL-2, IL-4, and IL-15. The receptor for IL-21 (IL-21R, also termed NILR for novel interleukin receptor) is a new member of the class I cytokine receptor family. IL-021R forms a complex with the common cytokine receptor g chain and mediates IL-21 signaling. IL-21 and its receptor activates JAK-STAT signaling pathway. IL-21 is expressed in activated T cells, HL-60, and THP-1 cell lines. IL-21 plays an important role in the proliferation and maturation of NK, B and T cell populations.

Recommended Dilutions: Western Blot: 1 µg/ml

Type: Primary
Antigen: IL21R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty