Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CTGF Rabbit Polyclonal Antibody

Supplier: BioVision

5553R-100
10007-822EA 272.95 USD InStock
10007-822
Anti-CTGF Rabbit Polyclonal Antibody
Antibodies
CTGF (connective tissue growth factor) is a cysteine-rich, secreted peptide. It is a new member of the peptide family that includes serum-induced immediate early gene products, a v-src-induced peptide, and a putative proto-oncogene. Among the many functions of the CTGF gene family are embryogenesis, wound healing, and regulation of extracellular matrix production.

  • Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunohistochemistry: 10-20 µg/ml, frozen and paraffin sections

Type: Primary
Antigen: CTGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty