Anti-BLC Rabbit Polyclonal Antibody

Supplier: BioVision
5550-100
10007-820EA 309.39 USD
10007-820
Anti-BLC Rabbit Polyclonal Antibody
BLC (B-lymphocyte chemoattractant) is a 12 kDa ELR-CXC chemokine. BLC showed high levels of expression in spleen, Peyer’s patches, and mesenteric lymph nodes. BLC is reported as the ligand of CXCR5. It attracts primarily B cells in vitro. Cell lines transfected with CXCR5 demonstrate both chemotaxis and calcium flux in response to BLC.

Recommended Dilutions: Western Blot: at 0.5-4 µg/ml

Type: Primary
Antigen: BLC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR