Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL19 Rabbit Polyclonal Antibody

Supplier: BioVision

5546-100
10008-090EA 252.35 USD InStock
10008-090
Anti-IL19 Rabbit Polyclonal Antibody
Antibodies
IL-19 is a single, non-glycosylated polypeptide with 152 amino acids. It plays an important role in regulating development and functioning of the skin .

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: IL19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty