Anti-C3AR Rabbit Polyclonal Antibody

Supplier: BioVision
5543-100
10008-088EA 309.39 USD
10008-088
Anti-C3AR Rabbit Polyclonal Antibody
C3a is an anaphylatoxin generated throμgh activation of the complement C3. C3a is a potent chemoattractant for phagocytes, and it stimulates chemotaxis and other leukocyte functions. C3a binds to eosinophils, basophils, neutrophils, and differentiated U937 cells, that all express receptors for C3a on their cell surface. The C3a receptor has been cloned and shown to be a G protein-coupled receptor with seven putative transmembrane domains. This receptor is characterized for its large extracellular loop between the fourth and fifth transmembrane domains. The loop can be readily detected using the rabbit anti-C3a receptor antibody.

Recommended Dilutions: 0

Type: Primary
Antigen: C3AR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR