Anti-CXCR4 Rabbit Polyclonal Antibody

Supplier: BioVision
5541-100
10008-082EA 309.39 USD
10008-082
Anti-CXCR4 Rabbit Polyclonal Antibody
CXCR4 (fusin) is a member in the seven-transmembrane domain G-protein-coupled chemokine receptor family. The full-length cDNA was isolated from a human spleen cDNA library. Its ligand is chemokine stroma-derived factor (SDF). CXCR4 has been shown to be the co-receptor for HIV-1 binding to CD4 throμgh the viral envelope glycoprotein gp120. In other cases, CXCR4 can function as the only receptor for the interaction of HIV-2 with CD4 and host cells. A mouse monoclonal antibody (12G5) to CXCR4 has been shown to inhibit HIV infectivity and HIV-induced syncytium.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CXCR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR