Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CX3CR1 Rabbit Polyclonal Antibody

Supplier: BioVision

5537R-100
10008-076EA 252.35 USD InStock
10008-076
Anti-CX3CR1 Rabbit Polyclonal Antibody
Antibodies
Human CX3CR1 has been shown to mediate both the adhesive and migratory functions of fractalkine. Fractalkine and CX3CR1 represent new types of leukocyte trafficking regulators.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CX3CR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty