Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SLC Rabbit Polyclonal Antibody

Supplier: BioVision

5534-100
10008-074EA 272.95 USD InStock
10008-074
Anti-SLC Rabbit Polyclonal Antibody
Antibodies
SLC (secondary lymphoid-tissue chemokine, also called 6Ckine, TCA4, Exodus-2) is a 111-amino acid (human; 110 a.a. for mouse) CC chemokine with an N-terminal sequence of 23 residues as a signal sequence. It was originally cloned in 1997. Recombinant SLC attracts T-, B- cells, mature dendritic cells, but did not induce any cell migration in neutrophils or monocytes. SLC is a functional ligand for CCR7.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml

Type: Primary
Antigen: SLC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty