Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MDC Rabbit Polyclonal Antibody

Supplier: BioVision

5531-100
10008-072EA 272.95 USD InStock
10008-072
Anti-MDC Rabbit Polyclonal Antibody
Antibodies
MDC (macrophage-derived chemokine) is a 69-amino acid CC chemokine with a N-terminal sequence of 24 residues as a signal sequence. It was originally cloned from human monocyte-derived macrophages. Recombinant MDC attracts monocyte-derived dendritic cells, activated lymphocytes and natural killer cells. MDC has anti-HIV-1 activity. Expression of MDC mRNA in macrophages is enhanced by LPS, IL-1β and TNFα. MDC is a functional ligand for CCR4.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: MDC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty