Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FABP4 Rabbit Polyclonal Antibody

Supplier: BioVision

5504-100
10008-064EA 252.35 USD InStock
10008-064
Anti-FABP4 Rabbit Polyclonal Antibody
Antibodies
Fatty Acid-binding Proteins (FABP) are a family of homologous 14-15 kDa cytoplasmic proteins that are expressed in highly tissue-specific manner and play an integral role in the balance between lipid and carbohydrate metabolism. FABPs have been reported to be involved in the uptake and metabolism of fatty acids, maintenance of cellular membrane fatty acids levels, intracellular trafficking, modulation of specific enzymes of lipid metabolic pathways, as well as in the modulation of cell growth and differentiation.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: FABP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty