Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Protein A Rabbit Polyclonal Antibody

Supplier: BioVision

5500-100
10007-818EA 262.65 USD InStock
10007-818
Anti-Protein A Rabbit Polyclonal Antibody
Antibodies
Protein A binds to the constant region of many species of immunoglobulin G. Protein A can be used to detect, quantify and purify IgG antibodies and antibody/antigen complexes. Protein A binds to all IgG subclasses from human and mouse species.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Protein A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty