Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PDGFB Rabbit Polyclonal Antibody

Supplier: BioVision

5489-100
10007-816EA 267.72 USD InStock
10007-816
Anti-PDGFB Rabbit Polyclonal Antibody
Antibodies
Mouse Platelet Derived Growth Factor (PDGF) is a potent mitogen for a wide range of cell types including fibroblasts, smooth muscle and connective tissue. PDGF-BB is a 24.4.0 kDa protein containing 109 amino acid residues. The reduced form of PDGF-BB shows ~14-18 kDa on SDS-PAGE.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: PDGFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty