Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MDK Rabbit Polyclonal Antibody

Supplier: BioVision

5479-100
10007-810EA 252.35 USD InStock
10007-810
Anti-MDK Rabbit Polyclonal Antibody
Antibodies
Human Midkine (MK) is a new member of the heparin-binding neurotrophic factor family. MK and PTN (Pleiotrophin) are structural homologs, and are highly conserved among species. MK plays important roles in development and carcinogenesis and has several important biological effects, including promotion of neurite extension and neuronal survival. Human midkine is a 13.4 kDa protein, comprising of 123 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-1 µ/ml; ELISA: 1-4 µg/ml

Type: Primary
Antigen: MDK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty