Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-OPN Mouse Monoclonal Antibody [clone: 2F10]

Supplier: BioVision

5423-100
10007-800EA 288.4 USD InStock
10007-800
Anti-OPN Mouse Monoclonal Antibody [clone: 2F10]
Antibodies
Human Osteopontin (OPN) is a negatively charged hydrophilic protein of 314 amino acids and is subject to significant post translational modifications (PTM) including phosphorylation and glycosylation. Due to its acidic nature and PTMs, OPN runs anomalously by SDS-PAGE. Althoμgh its mass is 35kD, apparent molecular weights may range up to 75kD. In addition, OPN is subject to proteolytic modification into smaller molecular weight fragments. The matricellular protein osteopontin binds to cell surface receptors and is secreted into many body fluids including milk, blood and urine, depending on the organ of origin. This makes osteopontin an ideal candidate for being a biomarker as the secreted form is easily obtained in throwaway fluids, and mimics the cellular environment from which it is released. Osteopontin is important in immune responses and inflammation as well as bone generation and remodeling. In autistic children, serum levels of osteopontin are correlated to the severity of disease, probably due to a brain inflammation pattern in these children. In aortic valve sclerosis and stenosis, increased levels of secreted osteopontin are also noted. Osteopontin has also been sμggested as a cancer biomarker, since it is associated with tumor formation, progression and metastasis. In bone and tooth formation osteopontin is known to be a negative regulator of parathyroid hormone-related protein receptor, which induces osteogenesis. Without appropriate levels of osteopontin, bone growth continues unregulated, and leads to specific bone cancers. In short, osteopontin is a strong marker for bone growth, inflammation and certain cancers. The newly exposed SVVYG epitope on the N-terminal fragment has also been shown to participate in cell adhesion.

Recommended Dilutions: 0

Type: Primary
Antigen: OPN
Clonality: Monoclonal
Clone: 2F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty