Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Nitrotyrosine Mouse Monoclonal Antibody [clone: HM11]

Supplier: BioVision

5414-50
10007-790EA 566.5 USD InStock
10007-790
Anti-Nitrotyrosine Mouse Monoclonal Antibody [clone: HM11]
Antibodies
Nitrotyrosine is a product of tyrosine nitration mediated by reactive nitrogen species such as peroxynitrite anion and nitrogen dioxide. It is a marker for inflammation and nitric oxide (NO) production. It is detected in large number of pathological conditions and is considered a marker of NO-dependent, reactive nitrogen species-induced nitrative stress. Tyrosine nitration can inactivate enzymes and receptors that depend on tyrosine residues for their activity. Nitration prevents phosphorylation of tyrosine residues important for signal transduction. Nitrotyrosine has been detected in inflammatory processes such as septic shock, rheumatoid arthritis, celiac disease, atherosclerotic plaques, chronic renal failure and keratoconus. Because nitrotyrosine is a stable product of multiple pathways, such as the formation of peroxynitrite, its plasma concentration may be a useful determinant of NO-dependent damage in vivo. Nitrotyrosine Antibody detects proteins and peptides containing nitro-tyrosine in a manner independent of the surrounding amino acid sequence. It is a valuable tool for identifying new nitrated proteins as well as for assaying protein nitration and measuring levels of nitrated proteins in tissues and samples.

Recommended Dilutions: Western Blot: 1 µg/ml) using secondary antibody coupled to alkaline phosphatase and BCIP/NBT as substrate, ELISA: 1:1000; Immunohistochemistry.

Type: Primary
Antigen: Nitrotyrosine
Clonality: Monoclonal
Clone: HM11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All Species

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty