Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-mCherry Mouse Monoclonal Antibody [clone: 1]

Supplier: BioVision

5411-100
10007-784EA 338.75 USD InStock
10007-784
Anti-mCherry Mouse Monoclonal Antibody [clone: 1]
Antibodies
mCherry is an engineered derivative of one of a family of proteins originally isolated from Cnidarians (jelly fish, sea anemones and corals). mCherry is the second generation monomeric red fluorescent protein that have improved brightness and photostabilty. It is an ideal tool for monitoring the dynamics of tagged proteins for protein-protein integration, cell-sorting, organelle labeling, protein localization, gene expression and more. The anti-mCherry antibody recognizes over expressed recombinant proteins containing the mCherry tag fused to either the amino- or carboxy termini of targeted proteins in transfected mammalian cells or other expression systems.

Recommended Dilutions: Western Blot: 10 µg/ml;

Type: Primary
Antigen: mCherry
Clonality: Monoclonal
Clone: 1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All Species

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty