Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SDF-1 beta Rabbit Polyclonal Antibody

Supplier: BioVision

5390A-100
10007-756EA 252.35 USD InStock
10007-756
Anti-SDF-1 beta Rabbit Polyclonal Antibody
Antibodies
Stromal cell-derived factor-1 beta is a stromal derived CXC chemokine that belong to the intercrine family, members of which activate leukocytes and are often induced by proinflammatory stimuli such as lipopolysaccharide, TNF, or IL-1. SDF-1β signals throμgh CXCR4 receptor.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SDF-1 beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty