Anti-SDF-1 beta Rabbit Polyclonal Antibody

Supplier: BioVision
5390-100
10008-002EA 286.04 USD
10008-002
Anti-SDF-1 beta Rabbit Polyclonal Antibody
Stromal cell-derived factor-1 beta is a stromal derived CXC chemokine that belong to the intercrine family, members of which activate leukocytes and are often induced by proinflammatory stimuli such as lipopolysaccharide, TNF, or IL1. SDF-1β signals throμgh CXCR4 receptor.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SDF-1 beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR