Anti-SDF-1 alpha Rabbit Polyclonal Antibody

Supplier: BioVision
5387-100
10008-000EA 297.63 USD
10008-000
Anti-SDF-1 alpha Rabbit Polyclonal Antibody
SDF-1α (stromal cell-derived factor-α), also known as pre-B-cell growth-stimulating factor (PBSF), is a 70-amino acid CXC chemokine originally cloned from a bone marrow stromal cell line. Targeted deletion of SDF-1 gene resulted in defects of B-cell lymphopoiesis and bone marrow myelopoiesis. SDF-1 has been shown to be chemotactic for lymphocytes. In addition, SDF-1 was recently reported to be a ligand for CXCR4 (LESTR/fusin), a co-receptor for HIV-1 entry into T cells. SDF-1 binding to CXCR4 inhibits HIV-1 entry.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml; Immunofluorescence/Neutralization: 2-10 µg/ml

Type: Primary
Antigen: SDF-1 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR