Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NP1 Goat Polyclonal Antibody

Supplier: BioVision

5384-100
10007-998EA 272.95 USD InStock
10007-998
Anti-NP1 Goat Polyclonal Antibody
Antibodies
Human alpha-defensin-1 (NP-1) is an antimicrobial peptide which belongs to the distinct family of alpha-defensins. Human NP-1 is a 3.4 kDa protein containing 30 amino acid residues.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml; ELISA: 2-4 µg/ml

Type: Primary
Antigen: NP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty