Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-XCL1 Rabbit Polyclonal Antibody

Supplier: BioVision

5376-100
10007-602EA 272.95 USD InStock
10007-602
Anti-XCL1 Rabbit Polyclonal Antibody
Antibodies
Lymphotactin is the only C chemokine so far identified which has a single cysteine residue near the amino terminus. It has 114 amino acids with a 22 amino acids signal peptide. It is produced in activated CD8+ T cells and NK cells. Lymphotactin attracts both T lymphocytes and NK cells.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 4-8 µg/ml

Type: Primary
Antigen: XCL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty