Anti-4-1BBL Rabbit Polyclonal Antibody

Supplier: BioVision
5369-100
10007-598EA 286.04 USD
10007-598
Anti-4-1BBL Rabbit Polyclonal Antibody
Human 4-1BB Ligand (4-1BBL) is a member of the emerging family of Ligands with structural homology to tumor necrosis factor. Human 4-1BBL is a 19.5 kDa protein containing 185 amino acid residues, comprising the TNF-like extra-cellular domain of 4-1BBL.

Recommended Dilutions: Western Blot: 0.5-4.0 µg/ml; Immunohistochemistry: 5 µg/ml

Type: Primary
Antigen: 4-1BBL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR