Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VEGF Rabbit Polyclonal Antibody

Supplier: BioVision

5363-100
10007-754EA 252.35 USD InStock
10007-754
Anti-VEGF Rabbit Polyclonal Antibody
Antibodies
VEGF (Vascular Endothelial Growth Factor) is a secreted homodimeric protein, secreted by a variety of vascularized tissues. VEGF stimulates endothelial cell growth, angiogenesis, and capillary permeability. Human VEGF is a 38.2 kDa homodimeric protein consisting of two 165 amino acid polypeptide chains.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml; ELISA: 2-3 µg/ml; Immunohistochemistry: 5 µg/ml; Neutralization: 5-10 µg/ml

Type: Primary
Antigen: VEGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty