Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNF-related Apoptosis-inducing Ligand Rabbit Polyclonal Antibody

Supplier: BioVision

5354-100
10007-752EA 252.35 USD InStock
10007-752
Anti-TNF-related Apoptosis-inducing Ligand Rabbit Polyclonal Antibody
Antibodies
Human TRAIL (TNF-Related Apoptosis Inducing Ligand), also called Apo2 Ligand (Apo2L), is a cytotoxic protein that activates rapid apoptosis in tumor cells, but not at normal cells. Human TRAIL/Apo2L is a single polypeptide chain containing 168 amino acids and has a molecular mass of 19.6 kDa.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TNF-related Apoptosis-inducing Ligand
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty