Anti-TPO Rabbit Polyclonal Antibody

Supplier: BioVision
5352-100
10007-750EA 286.04 USD
10007-750
Anti-TPO Rabbit Polyclonal Antibody
TPO (Thrombopoietin, also known as Mpl-ligand, MGDF) stimulates the proliferation and maturation of megakaryocytes and promotes increased circulation levels of platelets in vivo.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TPO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR